missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Galactose, >99%

Product Code. 15885138 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15885138 500 g Plastic bottle 500g
15835288 1 kg Plastic bottle 1kg
15839762 5 kg Plastic bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15885138 Supplier Thermo Scientific Supplier No. J15995.36
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59-23-4 | C6H12O6

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00151230

Specifications

Melting Point 164°C to 168°C
Density 1.5
Packaging Plastic bottle
Quantity 500 g
Beilstein 1724619
Specific Gravity 1.50
Formula Weight 180.16
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D-(+)-Galactose

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.