missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Galactose, 99+%

Product Code. 10307470 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
25 kg
Unit Size:
100g
1kg
25kg
500g
5kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10307470 500 g 500g
10011560 100 g 100g
10427360 1 kg 1kg
10649283 5 kg 5kg
15224644 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10307470 Supplier Thermo Scientific Acros Supplier No. 150615000
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D-Galactose, CAS # 59-23-4, is a right-handed conformation of a monosaccharide that is grouped as an aldohexose, and the molecule may be useful in study of aging, inflammation, and metabolic disorders. | CAS: 59-23-4 | C6H12O6

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

  • Galactose is a monosaccharide and aldohexose found in most organisms
  • Galactose is an epimer of glucose and can be converted to glucose through the Leloir pathway
  • It also plays a role as a metabolite in mice and E. coli
  • Galactose is less stable than glucose and rapid conversion often occurs
  • Galactose may also be incorporated into glycoconjugates

Applications

  • Metabolism of galactose may be relevant to the study of aging, inflammation, and metabolic disorders
  • It is intended for a variety of applications from in vitro metabolic assays to HPLC
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6H12O6
MDL Number MFCD00063833

Specifications

Melting Point 165.0°C to 168.0°C
Color White
Density 6.2
Infrared Spectrum Authentic
Assay Percent Range 99+%
Quantity 500 g
Beilstein 31, 295
Merck Index 15, 4365
Specific Rotation Condition + 79.75 (20.00°C c=10,H2O,NH3)
Specific Rotation + 79.75
Formula Weight 180.16
Vapor Pressure Negligible
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D(+)-Galactose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.