missing translation for 'onlineSavingsMsg'
Learn More

D-Galactose, TRC

Product Code. 30137102
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137102 500 g 500g
30005085 10 g 10g
30007090 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137102 Supplier Toronto Research Chemicals Supplier No. TRCG155250500G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-23-4
Molecular Formula C6 H12 O6
Molecular Weight (g/mol) 180.16
Synonym D-Galactose, Galactose, D- (8CI), (+)-Galactose, D-(+)-Galactose, Galactose, D-Galactopyranose, D(+)-Galactose
IUPAC Name (3R,4S,5R,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@H]1OC(O)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Quantity 500 g
InChI Formula InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3+,4+,5-,6?/m1/s1
Formula Weight 180.0634
Percent Purity >95%
Chemical Name or Material D-(+)-Galactose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.