missing translation for 'onlineSavingsMsg'
Learn More

D-Gluconic acid, 50% aq. soln.

Product Code. 11432838
Change view
Click to view available options
Quantity:
500 mL
2500 mL
Unit Size:
2500mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11432838 2500 mL 2500mL
11422838 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11432838 Supplier Thermo Scientific Alfa Aesar Supplier No. A16806.0F
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 526-95-4 | C6H12O7 | 196.155 g/mol

Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Gluconic acid occurs naturally in fruit, honey, and wine. As a food additive it is an acidity regulator. It is also used in cleaning products.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-95-4
Molecular Formula C6H12O7
Molecular Weight (g/mol) 196.155
MDL Number MFCD00004240
InChI Key RGHNJXZEOKUKBD-SQOUGZDYSA-N
Synonym gluconic acid, d-gluconic acid, dextronic acid, maltonic acid, glycogenic acid, glosanto, gluconate, pentahydroxycaproic acid, gluconic acid, d, d-gluconate
PubChem CID 10690
ChEBI CHEBI:33198
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid
SMILES C(C(C(C(C(C(=O)O)O)O)O)O)O

Specifications

Density 1.234
Quantity 2500 mL
Optical Rotation +10° (Neat)
Refractive Index 1.418
UN Number UN3265
Beilstein 1726055
Merck Index 144456
Solubility Information Soluble in water.
Formula Weight 196.16 (Anhydrous)
Concentration or Composition (by Analyte or Components) 50% aq. soln.
Chemical Name or Material D-Gluconic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.