missing translation for 'onlineSavingsMsg'
Learn More

D-Gluconic acid, calcium salt, 99%

Product Code. 10591651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Unit Size:
1kg
250g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10591651 250 g Plastic bottle 250g
10358250 1 kg Plastic bottle 1kg
10277263 5 kg Plastic drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10591651 Supplier Thermo Scientific Acros Supplier No. 211062500
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 299-28-5 | C12H22CaO14 | 430.372 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 299-28-5
Molecular Formula C12H22CaO14
Molecular Weight (g/mol) 430.372
MDL Number MFCD00064209
InChI Key NEEHYRZPVYRGPP-IYEMJOQQSA-L
Synonym calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
PubChem CID 9290
IUPAC Name calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
SMILES C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]

Specifications

Color White
Loss on Drying 2.0 % max. (1 g, 105°C, 16 hrs)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula (HOCH2[CH(OH)]4CO2)2Ca
Quantity 250 g
Beilstein 03, 544
Merck Index 15, 1671
Specific Rotation Condition + 10.20 (22.00°C c=1,H2O)
Specific Rotation + 10.20
Formula Weight 430.38
Percent Purity 0.99
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Gluconic acid, calcium salt
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.