missing translation for 'onlineSavingsMsg'
Learn More

D-Gluconic Acid Potassium Salt, TRC

Product Code. 30406019
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406019 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30406019 Supplier Toronto Research Chemicals Supplier No. TRCG417425100G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Gluconate
CAS 299-27-4
Molecular Formula C6 H11 O7 . K
Quantity 100 g
Synonym Potassium Gluconate
InChI Formula InChI=1S/C6H12O7.K/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1
SMILES [K+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C(=O)[O-]
IUPAC Name potassium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
Molecular Weight (g/mol) 234.25
Formula Weight 234.0142
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.