missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucono-1,5-lactone, MedChemExpress

Product Code. 16441238
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16441238 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16441238 Supplier Medchem Express Supplier No. HYI0301
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-(+)-Glucono-1,5-lactone is a polyhydroxy (PHA) that is capable of metal chelating, moisturizing and antioxidant activity.

D-(+)-Glucono-1,5-lactone is a polyhydroxy (PHA) that is capable of metal chelating, moisturizing and antioxidant activity.

Specifications

Chemical Name or Material D-(+)-Glucono-1,5-lactone
CAS 90-80-2
Color White
Purity Grade Notes Research
Molecular Formula C6H10O6
Quantity 5 g
Synonym Gluconic acid lactone
Solubility Information DMSO : 100 mg/mL (561.36 mM; Need ultrasonic) ∣H2O : ≥ 100 mg/mL (561.36 mM)
SMILES O=C1[C@H](O)[C@@H](O)[C@H](O)[C@@H](CO)O1
Molecular Weight (g/mol) 178.14
Formula Weight 178.14
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.