missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose Anhydrous, Extra Pure, SLR, Fisher Chemical™

Product Code. 10335850 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
2kg
500g
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10335850 500 g Plastic powder jar 500g
10395890 1 kg Plastic powder jar 1kg
10254480 2 kg Plastic powder jar 2kg
10539380 5 kg Plastic keg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10335850 Supplier Fisher Chemical Supplier No. G/0450/53
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

D(+)-Glucose Anhydrous, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
MDL Number 148912
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146°C
Color White
pH 6 to 7
Packaging Plastic powder jar
Quantity 500 g
Formula Weight 180.16g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.