missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucose, anhydrous, For ACS analysis, MP Biomedicals™

Product Code. 11482842 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
5 kg
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11482842 500 g 500g
11492842 5 kg 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11482842 Supplier MP Biomedicals Supplier No. 0215252790
  • ONLINE EXCLUSIVE PRICE
    49.85 €
    47.25 € / 500g
    Save 2.60 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.156
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Density 1.544g/mL
Quantity 500 g
Formula Weight 180.2
Physical Form Powder
Chemical Name or Material D-(+)-Glucose, For ACS analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.