missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP

Product Code. 15582943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15582943 1 kg 1kg
15541300 5 kg 5kg
15501391 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15582943 Supplier Thermo Scientific Acros Supplier No. 388190010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 50-99-7 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
InChI Key GZCGUPFRVQAUEE-SLPGGIOYSA-N
Synonym 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, d +-glucose, aldehydo-d-glucose, glucose, anhydrous, dextrose, anhydrous, d-glucose, anhydrous, glucosteril, anhydrous dextrose, dextrose solution, d-glucose in linear form
PubChem CID 107526
ChEBI CHEBI:42758
IUPAC Name (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 146.0°C
Loss on Drying 0.5% max. (1 g, 105°C, 16 h)
Quantity 1 kg
Merck Index 15,4495
Specific Rotation Condition + 52.75 (25.00°C c=10, H2O, NH4OH)
Specific Rotation + 52.75
Solubility Information Solubility in water: soluble.
Formula Weight 180.16
Percent Purity 97.5 to 102.0% (Anhydrous substance)
Related Substances 0.5% max. (total impurities)
Chemical Name or Material D(+)-Glucose, anhydrous, specificied according to the requirements of Ph.Eur., USP, BP
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.