missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Glucose, MP Biomedicals™

Product Code. 11485172 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485172 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11485172 Supplier MP Biomedicals Supplier No. 02194672.1
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D-(+)-Glucose Anhydrous, Cell Culture Reagent, White Powder

  • D-(+)-Glucose, anhydrous is a main source of energy for living organisms.
  • It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen.
  • D-(+)-Glucose is a monosaccharide. It is a mixture of α- and β-anomers, primarily the α-anomer.

Specifications

CAS 50-99-7
Melting Point 150°C to 170°C (literature)
Color White
Density 1.544g/mL
Molecular Formula C6H12O6
Quantity 100 g
Beilstein 1724615
Synonym Dextrose, Cartose, Cerelose, Corn Sugar
Solubility Information Soluble in 20% aqueous clear colorless solution.
InChI Key WQZGKKKJIJFFOK-UHFFFAOYNA-N
Optical Rotation (c=10, Water, NH4OH) - +52.5 ± 5°
Molecular Weight (g/mol) 180.16
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.