missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- β-D-Glucose is a main source of energy for living organisms
- It is combined into glucosides, disaccharides, oligosaccharides, the polysaccharides (cellulose and starch), and glycogen
Specifications
Specifications
| Melting Point | 148°C to 155°C (literature) |
| Color | White |
| Boiling Point | ∼410.8°C (760 mmHg (Predicted) (literature)) |
| Linear Formula | C6H12O6 |
| Quantity | 100 g |
| Solubility Information | Soluble in water (1/20-clear solution) and alcohol (slightly). |
| Formula Weight | 180.16 |
| Percent Purity | ≥80% |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | β-D-GLUCOSE |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?