missing translation for 'onlineSavingsMsg'
Learn More

D-Glucose, TRC

Product Code. 30005976
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005976 250 g 250g
30006152 100 g 100g
17969974 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005976 Supplier Toronto Research Chemicals Supplier No. TRCG595000250G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-99-7
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
Synonym (+)-Glucose, Anhydrous dextrose, Brake (pH-encapsulated glucose), C*Dry GL 01934, CPC hydrate, Cartose, Cerelose, Cerelose 2001, Clearsweet 95, Clintose L, Corn sugar, D 50W, D(+)-Glucose, Dextropur, Dextrose, Dextrosol, FeedBead, Glucodin, Glucolin, Glucopyranose, Glucose, Glucosoft, Glucosteril, Goldsugar, Grape sugar, Hi-Mesh, Maxim Energy Gel, Meritose, Meritose 200, NSC 287045, Roclys C 30725, Roferose ST, Staleydex 111, Staleydex 130, Staleydex 333, Staleydex 95M, Sugar, grape, TDA-C, TDA-S, Tabfine 097(HS), Tackidex 30L75, Toleran G, Vadex
IUPAC Name (2RS,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydropyran-2,3,4,5-tetrol
SMILES O[C@H]([C@@H](CO)OC(O)[C@@H]1O)[C@@H]1O

Specifications

Quantity 250 g
InChI Formula InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6?/m1/s1
Formula Weight 180.06
Percent Purity >95%
Chemical Name or Material Dextrose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.