missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucuronic acid, sodium salt, monohydrate, 98%, Thermo Scientific Chemicals

Product Code. 10226740 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10226740 25 g Plastic bottle 25g
10627304 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10226740 Supplier Thermo Scientific Acros Supplier No. 204570250
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 14984-34-0 | C6H9NaO7·H2O | 234.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14984-34-0
Molecular Formula C6H9NaO7·H2O
Molecular Weight (g/mol) 234.14
MDL Number MFCD00151051
InChI Key QKHMTHNLNZGTSP-JSCKKFHOSA-N
Synonym sodium d-glucuronate
PubChem CID 87235405
IUPAC Name sodium;(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
SMILES C(=O)C(C(C(C(C(=O)O)O)O)O)O.[Na]

Specifications

Melting Point 134°C to 140°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 25 g
Beilstein 03, 886
Merck Index 10, 4327
Specific Rotation Condition +21° (20°C c=2,H2O,after 4hrs.)
Specific Rotation + 21
Formula Weight 234.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Glucuronic acid, sodium salt, monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.