missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucurono-3,6-lactone, 99+%

Product Code. 10104710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10104710 500 g Plastic bottle 500g
10583581 25 g Plastic bottle 25g
10388450 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10104710 Supplier Thermo Scientific Acros Supplier No. 225925000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32449-92-6 | C6H8O6 | 176.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32449-92-6
MDL Number MFCD00135622
InChI Key UYUXSRADSPPKRZ-KKQCNMDGSA-N
Synonym d-glucuro-3,6-lactone
PubChem CID 91634076
IUPAC Name (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
SMILES C(=O)C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 170.0°C to 176.0°C
Color White
Loss on Drying 0.5% max. (80°C)
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C6H8O6
Quantity 500 g
Merck Index 15, 4503
Specific Rotation Condition +19.00 (20.00°C c=10,H2O)
Specific Rotation + 19.00
Solubility Information Solubility in water: soluble in water. Other solubilities: slightly soluble in glacial acetic acid, slightly soluble in methanol and absolute, ethanol
Molecular Weight (g/mol) 176.124
Formula Weight 176.12
Percent Purity ≥99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D(+)-Glucurono-3, 6-lactone, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.