missing translation for 'onlineSavingsMsg'
Learn More

D-Glucurono-3,6-lactone, TRC

Product Code. 30134338
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134338 25 g 25g
30133783 5 g 5g
30007724 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134338 Supplier Toronto Research Chemicals Supplier No. TRCG59690025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32449-92-6
Molecular Formula C6 H8 O6
Molecular Weight (g/mol) 176.12
Synonym D-Glucuronolactone, D-Glucuronic acid, γ-lactone, Glucuronic acid, γ-lactone, D- (6CI,8CI), D-Glucofuranurono-6,3-lactone, D-Glucuronic acid lactone, D-Glucuronic acid γ-lactone, D-Glucurono-3,6-lactone, D-Glucurono-6,3-lactone, D-Glucurono-γ-lactone, D-Glucuronolactone, Dicurone, Glucoxy, Glucurolactone, Glucurone, Glucuronic acid γ-lactone, Glucurono-6,3-lactone, Glucuronolactone, Glucuronosan, Glycurone, Guronsan, NSC 656, Reulatt S.S.
IUPAC Name (2R)-2-[(2S,3R,4S)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
SMILES O[C@@H](C=O)[C@H]1OC(=O)[C@@H](O)[C@H]1O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H8O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h1-5,8-10H/t2-,3+,4-,5+/m0/s1
Formula Weight 176.0321
Percent Purity >95%
Chemical Name or Material Glucuronolactone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.