missing translation for 'onlineSavingsMsg'
Learn More

D-Glucurono-6,3-lactone, 99%

Product Code. 11442558
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11442558 25 g 25g
11452558 100 g 100g
11462558 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11442558 Supplier Thermo Scientific Alfa Aesar Supplier No. A15861.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 32449-92-6 | C6H8O6 | 176.124 g/mol

D-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Glucurono-6,3-lactone is a glucuronic acid derivative studied for its effectiveness against canine hepatitis. It is utilized in the studies such as starting regent in the synthesis of 2,3,4,-tris(tert.-butyldimethysilyl) glucuronic acid trichloroethylester, required for the preparation of 1-O-acyl glucuronide of the anti-inflammatory drug ML-3000, synthesis of optically active glucopyranoses, synthesis of long-chain alkyl glucofuranosides.

Solubility
Soluble in Water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong bases, strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32449-92-6
Molecular Formula C6H8O6
Molecular Weight (g/mol) 176.124
MDL Number MFCD00135622
InChI Key UYUXSRADSPPKRZ-KKQCNMDGSA-N
Synonym d-glucuro-3,6-lactone
PubChem CID 91634076
IUPAC Name (2S)-2-[(2S,3S,4R)-3,4-dihydroxy-5-oxooxolan-2-yl]-2-hydroxyacetaldehyde
SMILES C(=O)C(C1C(C(C(=O)O1)O)O)O

Specifications

Melting Point 174°C (decomposition)
Quantity 25 g
Beilstein 83595
Merck Index 14,4467
Solubility Information Soluble in Water.
Optical Rotation +19° (c=8 in Water)
Formula Weight 176.12
Percent Purity 99%
Chemical Name or Material D-Glucurono-6,3-lactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.