missing translation for 'onlineSavingsMsg'
Learn More

D-Glutamic acid dimethyl ester hydrochloride, 98%

Product Code. 15468708
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468708 5 g 5g
15458708 1 g 1g
15437076 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15468708 Supplier Thermo Scientific Alfa Aesar Supplier No. H62132.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 27025-25-8 | C7H14ClNO4 | 211.642 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27025-25-8
Molecular Formula C7H14ClNO4
Molecular Weight (g/mol) 211.642
MDL Number MFCD00235872
InChI Key MFUPLHQOVIUESQ-NUBCRITNSA-N
Synonym dimethyl d-glutamate hydrochloride, h-d-glu ome-ome.hcl, h-d-glu ome-ome hcl, r-dimethyl 2-aminopentanedioate hydrochloride, unii-1jdm33eygh, d-glutamic acid, dimethyl ester, hydrochloride, 1jdm33eygh, d-glutamic acid dimethyl ester hydrochloride, d-glutamic acid 1,5-dimethyl ester hydrochloride, 1,5-dimethyl 2r-2-aminopentanedioate hydrochloride
PubChem CID 12917567
IUPAC Name dimethyl (2R)-2-aminopentanedioate;hydrochloride
SMILES COC(=O)CCC(C(=O)OC)N.Cl

Specifications

Melting Point 91°C
Quantity 5 g
Formula Weight 211.64
Optical Rotation −26° (c=5 in Water)
Sensitivity Hygroscopic
Percent Purity 98%
Chemical Name or Material D-Glutamic acid dimethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.