missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glutamine, 99+%

Product Code. 15493489
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493489 25 g 25g
15473489 1 g 1g
15483489 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493489 Supplier Thermo Scientific Chemicals Supplier No. J60784.14
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Kinase inhibitor; has antivasospastic properties | CAS: 5959-95-5 | C5H10N2O3 | 146.146 g/mol

Kinase inhibitor; has antivasospastic properties

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5959-95-5
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00065607
InChI Key ZDXPYRJPNDTMRX-GSVOUGTGSA-N
Synonym d-glutamine, h-d-gln-oh, d-2-aminoglutaramic acid, l +-glutamine, 2r-2-amino-4-carbamoylbutanoic acid, r-2,5-diamino-5-oxopentanoic acid, d-glutamic acid 5-amide, d-glutamin, unii-63hb36ca2y, d-gln
PubChem CID 145815
ChEBI CHEBI:17061
IUPAC Name (2R)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Color White
Beilstein 1723796
Quantity 25 g
Formula Weight 146.14
Percent Purity ≥99%
Physical Form Powder
Chemical Name or Material D-Glutamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.