missing translation for 'onlineSavingsMsg'
Learn More

D-Gulonic acid-1,4-lactone, 98%

Product Code. 15428579
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428579 5 g 5g
15438579 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428579 Supplier Thermo Scientific Alfa Aesar Supplier No. J66987.06
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6322-07-2 | C6H10O6 | 178.14 g/mol

D-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Gulono-1,4-lactone is a hexanoic acid used as a starting material in EPC syntheses.

Solubility
Soluble in water

Notes
Store at room temperature. Incompatible with heat, oxidizing agents. Ensure good ventilation.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6322-07-2
MDL Number MFCD00005391

Specifications

Melting Point 182°C to 188°C
Color White
Molecular Formula C5H2Br2O2S
Quantity 5 g
Beilstein 83012
Synonym 4,5-dibromo-2-thiophenecarboxylic acid, 4,5-dibromothenoic acid, 4,5-dibromo-thiophene-2-carboxylic acid, pubchem7747, acmc-20a4op, maybridge1_002274, 4-dimethylaminoethyl benzoate, 4,5-dibromo-2-thiophencarboxylic acid, 2,3-dibromothiophene-5-carboxylic acid, 2-thiophenecarboxylicacid, 4,5-dibromo
Solubility Information Soluble in water
InChI Key USFXRYVNRUMABJ-UHFFFAOYSA-N
SMILES C1=C(SC(=C1Br)Br)C(=O)O
IUPAC Name 4,5-dibromothiophene-2-carboxylic acid
Molecular Weight (g/mol) 285.94
PubChem CID 80611
Formula Weight 178.14
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Gulonic acid-1,4-lactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.