missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Gulonic acid gamma-lactone, 99%

Product Code. 10358100 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10358100 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10358100 Supplier Thermo Scientific Supplier No. 204450250
 Restricted Item
Estimated Shipment: 05-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6322-07-2 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 6322-07-2
Melting Point 182°C to 188°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C6H10O6
MDL Number MFCD00005391
Quantity 25 g
Beilstein 18, 204
Specific Rotation Condition −54.50° (20°C c=4,water)
Specific Rotation -54.5
InChI Key SXZYCXMUPBBULW-UHFFFAOYNA-N
SMILES OCC(O)C1OC(=O)C(O)C1O
IUPAC Name 5-(1,2-dihydroxyethyl)-3,4-dihydroxyoxolan-2-one
Molecular Weight (g/mol) 178.14
Formula Weight 178.14
Percent Purity 99%
Physical Form Crystals or Powder
Chemical Name or Material D(-)-Gulonic acid γ-lactone, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.