missing translation for 'onlineSavingsMsg'
Learn More

(D-His2)-Leuprolide Trifluoroacetic Acid Salt, TRC

Product Code. 30133315
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
100mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133315 250 mg 250mg
30137262 100 mg 100mg
30009334 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133315 Supplier Toronto Research Chemicals Supplier No. TRCL330610250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112642-11-2
Synonym 2-D-Histidine-6-D-leucine-9-(N-ethyl-L-prolinamide)-1-9-luteinizing Hormone-releasing Factor (swine) 2,2,2-Trifluoroacetate∣2: PN: WO2009040073 PAGE: 98 Claimed Protein;

Specifications

Format Neat
Molecular Formula C59H84N16O12
MDL Number MFCD02264520
Molecular Weight (g/mol) 1209.398, 1209.42
InChI Key GFIJNRVAKGFPGQ-JFMQHNAUSA-N
SMILES CCNC(=O)[C@@H]1CCCN1C(=O)[C@H](CCCN=C(N)N)NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](CC(C)C)NC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)[C@H](CO)NC(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)[C@@H](CC1=CN=CN1)NC(=O)[C@@H]1CCC(=O)N1
IUPAC Name (2S)-1-[(2S)-5-[(diaminomethylidene)amino]-2-[(2S)-2-[(2R)-2-[(2S)-2-[(2S)-3-hydroxy-2-[(2S)-2-[(2R)-3-(1H-imidazol-5-yl)-2-{[(2S)-5-oxopyrrolidin-2-yl]formamido}propanamido]-3-(1H-indol-3-yl)propanamido]propanamido]-3-(4-hydroxyphenyl)propanamido]-4-methylpentanamido]-4-methylpentanamido]pentanoyl]-N-ethylpyrrolidine-2-carboxamide
Quantity 250 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.