missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-Isoascorbic acid, 98%

Product Code. 11357808
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11357808 100 g 100g
11367808 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357808 Supplier Thermo Scientific Alfa Aesar Supplier No. 036366.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol

D-(-)-Isoascorbic acid is used as antioxidant especially in brewing industry, reducing agent in photography. And it is also used in food industry, as food additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-65-6
Molecular Formula C6H7NaO6
Molecular Weight (g/mol) 198.11
MDL Number MFCD00005378
InChI Key IFVCRSPJFHGFCG-HXPAKLQESA-N
Synonym erythorbic acid, isoascorbic acid, d-araboascorbic acid, d-isoascorbic acid, araboascorbic acid, d-erythorbic acid, isovitamin c, neo-cebicure, saccharosonic acid, mercate 5
PubChem CID 54675810
ChEBI CHEBI:51438
IUPAC Name (2R)-2-[(1R)-1,2-dihydroxyethyl]-3,4-dihydroxy-2H-furan-5-one
SMILES [Na+].OC[C@H](O)C1OC(=O)[C-](O)C1=O

Specifications

Melting Point 169°C to 172°C (decomposition)
Odor Odorless
Quantity 100 g
Merck Index 14,5126
Solubility Information Soluble in alcohol,pyridine and water.
Formula Weight 176.12
Percent Purity 99%
Physical Form Powder
Chemical Name or Material D-(-)-Isoascorbic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.