missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Isoleucine, 98%

Product Code. 11461720
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11461720 1 g 1g
15443697 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11461720 Supplier Thermo Scientific Chemicals Supplier No. H27488.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 319-78-8 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 319-78-8
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00064221
InChI Key AGPKZVBTJJNPAG-LXDRQGDMNA-N
Synonym d-isoleucine, 2r,3r-2-amino-3-methylpentanoic acid, h-d-ile-oh, isoleucine, d, unii-v87gja0g54, r-isoleucine, 2r,3r-2-amino-3-methylvaleric acid, alloisoleucin, d-ile, ambotzhaa1186
PubChem CID 76551
ChEBI CHEBI:27730
IUPAC Name (2R,3R)-2-amino-3-methylpentanoic acid
SMILES CC[C@@H](C)[C@@H](N)C(O)=O

Specifications

Melting Point 265°C (sublimation)
Quantity 1 g
Formula Weight 131.18
Optical Rotation −40° (c=5 in 5N HCl)
Percent Purity 98%
Chemical Name or Material D-Isoleucine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.