missing translation for 'onlineSavingsMsg'
Learn More

D,L-Alpha-Hydroxy Myristic Acid, TRC

Product Code. 30406511
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30406511 10 mg -
30398796 50 mg 50mg
30393527 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406511 Supplier Toronto Research Chemicals Supplier No. TRCH94850010MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2507-55-3
Molecular Formula C14 H28 O3
Molecular Weight (g/mol) 244.37
Synonym D,L-α-Hydroxy Myristic Acid
IUPAC Name 2-hydroxytetradecanoic acid
SMILES CCCCCCCCCCCCC(O)C(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H28O3/c1-2-3-4-5-6-7-8-9-10-11-12-13(15)14(16)17/h13,15H,2-12H2,1H3,(H,16,17)
Formula Weight 244.2038
Chemical Name or Material D,L-a-Hydroxymyristic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.