missing translation for 'onlineSavingsMsg'
Learn More

D,L-Venlafaxine-d6 (100 μg/mL in Methanol), TRC

Product Code. 30391109
Change view
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30391109 5 x 1 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30391109 Supplier Toronto Research Chemicals Supplier No. TRCKIT13575X1ML
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1020720-02-8
Molecular Formula C17H21D6NO2
Quantity 5 x 1 mL
Synonym 1-[2-(Dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexanol-d6
InChI Formula InChI=1S/C17H27NO2/c1-18(2)13-16(17(19)11-5-4-6-12-17)14-7-9-15(20-3)10-8-14/h7-10,16,19H,4-6,11-13H2,1-3H3/i4D2,5D2,6D2
SMILES OC1(CC([2H])([2H])C([2H])([2H])C([2H])([2H])C1)C(CN(C)C)C2=CC=C(OC)C=C2
IUPAC Name 3,3,4,4,5,5-hexadeuterio-1-[2-(dimethylamino)-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol
Molecular Weight (g/mol) 283.44
Formula Weight 283.2418
Concentration or Composition (by Analyte or Components) D,L-Venlafaxine-d6 100 μg/mL
Chemical Name or Material D,L-Venlafaxine-d6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.