missing translation for 'onlineSavingsMsg'
Learn More

D-Lactic acid

Product Code. p-7042252
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15454359 250 mg 250mg
15444359 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15454359 Supplier Thermo Scientific Alfa Aesar Supplier No. J61871.MD
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Useful chiral synthon, building block for depsipeptides | CAS: 10326-41-7 | C3H6O3 | 90.078 g/mol

D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(-)-Lactic acid is a useful biochemical for proteomics research. It is also used in the production of chiral drug and in the chemical synthesis. Lactic acid is used as a reagent in organic synthesis (in the manufacture of adhesives). It is used in the leather, textile, and tanning industries. It may be used as a plasticizer, a catalyst, or an acidifying agent. Lactic acid has even been used as a flavoring agent in the manufacture of tobacco products.

Solubility
Soluble in water (50mg/ml)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature -20°C. Keep away from strong oxidizing agents, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-41-7
Molecular Formula C3H6O3
Molecular Weight (g/mol) 90.078
MDL Number MFCD00068313
InChI Key JVTAAEKCZFNVCJ-UWTATZPHSA-N
Synonym d-lactic acid, r-2-hydroxypropanoic acid, r-lactic acid, --lactic acid, 2r-2-hydroxypropanoic acid, d-lactate, r---lactic acid, r-lactate, propanoic acid, 2-hydroxy-, 2r, d-milchsaeure
PubChem CID 61503
ChEBI CHEBI:42111
IUPAC Name (2R)-2-hydroxypropanoic acid
SMILES CC(C(=O)O)O

Specifications

Color White
Beilstein 1720254
Merck Index 14,5335
Solubility Information Soluble in water (50mg/ml)
Formula Weight 90.08
Quantity 250 mg
Optical Rotation −0.4° (c=8 in Water)
Physical Form Low Melting Solid
Chemical Name or Material D-Lactic acid
compliance-icons
Product Identifier
  • D-Lactic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: (R)-Lactic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.