missing translation for 'onlineSavingsMsg'
Learn More

D-Lactic Acid, TRC

Product Code. 30464788
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464788 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464788 Supplier Toronto Research Chemicals Supplier No. TRCL113485100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-Lactic Acid
CAS 10326-41-7
Molecular Formula C3H6O3
Quantity 100 g
Synonym (-)-Lactic Acid, (2R)-2-Hydroxypropanoic Acid, (R)-(-)-Lactic Acid, (R)-2-Hydroxypropanoic Acid, (R)-2-Hydroxypropionic Acid, (R)-Lactic Acid, (R)-α-Hydroxypropionic Acid, D-(-)-Lactic Acid, D-Lactic Acid, l-Lactic Acid
InChI Formula InChI=1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m1/s1
SMILES O[C@@H](C)C(O)=O
IUPAC Name (2R)-2-hydroxypropanoic acid
Molecular Weight (g/mol) 90.08
Formula Weight 90.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.