missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals alpha-D-Lactose monohydrate, 99.5+%

Product Code. 10501181 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
2 kg
5 kg
Packaging:
Glass bottle
Plastic Bottle
Plastic Drum
Unit Size:
1g
2kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10501181 1 g Glass bottle 1g
10665362 2 kg Plastic Bottle 2kg
10187703 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10501181 Supplier Thermo Scientific Chemicals Supplier No. 125090010
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

alpha-D-Lactose monohydrate, synonym lactose, CAS # 5989-81-1, is the hydrate form of the disaccharide lactose. | CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Lactose is a disaccharide composed of glucose and galactose
  • Lactose is the major sugar found in dairy products such as milk and cheese
  • alpha-D-Lactose monohydrate is used as a carrier and stabilizer of products in the pharmaceutical and food industries

Applications

  • Lactose has been used in vitro to interact with carbohydrate recognition domains on a recombinant form of LGALS9, thus blocking downstream functions of the protein
  • It has been used as a control to study sugar dependency in Drosophila, which do not digest lactose
  • As a common component of pharmaceutical tablets, alpha-lactose has been used in laboratory settings to imitate drug administration
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material α-D-Lactose monohydrate
Acidity 0.4mL 0.1N NaOH/6g max.
CAS 5989-81-1, 63-42-3
Absorbance Passes Test
CAS Min % 99.5
CAS Max % 100.0
Color White
Loss on Drying 0.5% max. (80°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 99.5+%
Packaging Glass bottle
Molecular Formula C12H24O12
MDL Number MFCD00150747
Quantity 1 g
Beilstein 31, 408
Merck Index 14, 5343
Specific Rotation Condition +54.40 (20.00°C c=10,H2O,NH3)
Specific Rotation +54.4° to +55.9° (20°C, 589nm) (c=10, H2O/NH3) on dry substance
Synonym alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x
InChI Key WSVLPVUVIUVCRA-KPKNDVKVSA-N
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
Molecular Weight (g/mol) 360.312
PubChem CID 104938
Formula Weight 360.3
Percent Purity ≥99.50%
Physical Form Crystalline Powder
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.