missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Lactose monohydrate, USP/NF, Ph.Eur., JP

Product Code. 10561892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10561892 1 kg Plastic bottle 1kg
10282812 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10561892 Supplier Thermo Scientific Acros Supplier No. 368890010
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

alpha-D-Lactose monohydrate, synonym lactose, CAS # 5989-81-1, is the hydrate form of the disaccharide lactose. | CAS: 5989-81-1 | C12H24O12 | 360.312 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Lactose is a disaccharide composed of glucose and galactose
  • Lactose is the major sugar found in dairy products such as milk and cheese
  • alpha-D-Lactose monohydrate is used as a carrier and stabilizer of products in the pharmaceutical and food industries

Applications

  • Lactose has been used in vitro to interact with carbohydrate recognition domains on a recombinant form of LGALS9, thus blocking downstream functions of the protein
  • It has been used as a control to study sugar dependency in Drosophila, which do not digest lactose
  • As a common component of pharmaceutical tablets, alpha-lactose has been used in laboratory settings to imitate drug administration
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5989-81-1
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.312
InChI Key WSVLPVUVIUVCRA-KPKNDVKVSA-N
Synonym alpha-d-lactose monohydrate, alpha-lactose monohydrate, lactose monohydrate, lactose, monohydrate, unii-ewq57q8i5x, respitose, alpha-lactose hydrate, a-lactose monohydrate, lactose monohydrate nf, ewq57q8i5x
PubChem CID 104938
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6S)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxyoxane-3,4,5-triol;hydrate
SMILES C(C1C(C(C(C(O1)OC2C(OC(C(C2O)O)O)CO)O)O)O)O.O

Specifications

Packaging Plastic bottle
Quantity 1 kg
Formula Weight 360.3
Chemical Name or Material α-D-Lactose monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.