missing translation for 'onlineSavingsMsg'
Learn More

D(-)-Leucinol, 97%, Thermo Scientific Chemicals

Product Code. 10269600 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10269600 1 g Glass bottle 1g
10678234 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10269600 Supplier Thermo Scientific Acros Supplier No. 413120010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53448-09-2
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00004734
InChI Key VPSSPAXIFBTOHY-ZCFIWIBFSA-O
Synonym d-leucinol, r-2-amino-4-methylpentan-1-ol, 2r-2-amino-4-methylpentan-1-ol, d---leucinol, chembl71171, r---leucinol, 1-pentanol, 2-amino-4-methyl-, 2r, 2-amino-4-methyl-pentan-1-ol, 2-amino-4-methyl-1-pentanol #
PubChem CID 2724002
IUPAC Name (2R)-2-amino-4-methylpentan-1-ol
SMILES CC(C)C[C@@H]([NH3+])CO

Specifications

Density 0.9070g/mL
Boiling Point 199.0°C
Flash Point 90°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4490 to 1.4530
Quantity 1 g
Specific Gravity 0.907
Specific Rotation Condition − 4.00 (20.00°C c=9,ethanol)
Specific Rotation − 4.00
Formula Weight 117.19
Percent Purity 97%
Chemical Name or Material D(-)-Leucinol, 97%, (97% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.