missing translation for 'onlineSavingsMsg'
Learn More

D-Limonene, 96.9%, MP Biomedicals

Product Code. 11463422 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463422 500 mL 500mL
11453422 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463422 Supplier MP Biomedicals Supplier No. 0215523490
  • ONLINE EXCLUSIVE PRICE
    103.00 €
    97.65 € / 500mL
    Save 5.35 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5989-27-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00062991
InChI Key XMGQYMWWDOXHJM-JTQLQIEISA-N
Synonym d-limonene, +-limonene, r-+-limonene, +-4r-limonene, +-carvene, +-dipentene, citrene, +-p-mentha-1,8-diene, 4r-limonene, d-+-limonene
PubChem CID 440917
ChEBI CHEBI:15382
SMILES CC(=C)[C@@H]1CCC(C)=CC1

Specifications

Color Colorless to Yellow
Quantity 500 mL
IUPAC Name (4R)-1-methyl-4-prop-1-en-2-ylcyclohexene
Formula Weight 136.26
Percent Purity 0.969
Physical Form Liquid
Chemical Name or Material D-Limonene
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • D-Limonene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P321-Specific treatment (see on this label).
  • P331-Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.