missing translation for 'onlineSavingsMsg'
Learn More

Melford D-Luciferin Free Acid

Product Code. p-200086991
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
This item is not returnable. View return policy

Product Code. 30018432

Brand: Melford L370000.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used with firefly luciferase in bioluminescence for the determination of ATP.

Used with firefly luciferase in bioluminescence for the determination of ATP.

Specifications

CAS 2591-17-5
Molecular Formula C11H8N2O3S2
Quantity 50 mg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.