missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress D-Luciferin sodium

Product Code. 16451467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451467 Supplier MedChemExpress Supplier No. HY12591
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Luciferin (D-(-)-Luciferin) sodium is the substrate of luciferases that catalyze the production of light in bioluminescent insects.

Specifications

CAS 103404-75-7
Solubility H2O : 250 mg/mL (826.99 mM∣Need ultrasonic) ∣DMSO : 100 mg/mL (330.80 mM∣Need ultrasonic)
Synonym D-(-)-Luciferin sodium∣Firefly luciferin sodium∣Beetle Luciferin sodium
Content And Storage 4°C, sealed storage, away from moisture and light∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture and light)
Quantity 50 mg
Molecular Weight (g/mol) 302.3 g/mol
Product Type Dye Reagents
Purity or Quality Grade 98.0%
Shipping Condition Room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.