Learn More
Melford D-Luciferin Sodium Salt
Product Code. 30018445
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
50mg
Product Code. 30018445
Brand: Melford L370800.25
RPI offers the highest purity D-Luciferin Sodium Salt for a wide range of bioluminescent applications.
RPI offers the highest purity D-Luciferin Sodium Salt for a wide range of bioluminescent applications. Allows highly sensitive bioluminescent measurements. Ideal for reporter gene assays, ATP assays, whole animal studied, DNA sequencing and more. Our product is specially purified and tested extensively to ensure the highest signal and reproducibility with low background and sensitivity. Guaranteed purity is determined by both standard HPLC and Chiral HPLC methods. Protect from light and moisture.
Specifications
Quantity 250 mg