missing translation for 'onlineSavingsMsg'
Learn More

Melford D-Luciferin Sodium Salt

Product Code. p-200086983
Click to view available options
Quantity:
50 mg
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
50mg
This item is not returnable. View return policy

Product Code. 30018446

Brand: Melford L370801.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

RPI offers the highest purity D-Luciferin Sodium Salt for a wide range of bioluminescent applications.

RPI offers the highest purity D-Luciferin Sodium Salt for a wide range of bioluminescent applications. Allows highly sensitive bioluminescent measurements. Ideal for reporter gene assays, ATP assays, whole animal studied, DNA sequencing and more. Our product is specially purified and tested extensively to ensure the highest signal and reproducibility with low background and sensitivity. Guaranteed purity is determined by both standard HPLC and Chiral HPLC methods. Protect from light and moisture.

Specifications

Quantity 1 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.