missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(-)-Lyxose, 99+%, mixture of anomers

Product Code. 10521271 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10521271 5 g Glass bottle 5g
10404225 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10521271 Supplier Thermo Scientific Chemicals Supplier No. 205230050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1114-34-7 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1114-34-7
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00064362
InChI Key PYMYPHUHKUWMLA-UOWFLXDJSA-N
Synonym 2s,3s,4r-2,3,4,5-tetrahydroxypentanal, d--lyxose, d-lyxo-pentose, unii-m795k6hjol, aldehydo-d-lyxose, m795k6hjol, dl-lyxose, +--lyxose, d-lyxose 9ci
PubChem CID 65550
ChEBI CHEBI:16789
IUPAC Name (2S,3S,4R)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 108.0°C to 112.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 31, 56
Merck Index 15, 5702
Specific Rotation Condition − 14.00 (20.00°C c=4,H2O,NH3)
Specific Rotation − 14.00
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in ethanol
Formula Weight 150.13
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material D(-)-Lyxose, Mixture of anomers
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.