missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Malic Acid, TRC

Product Code. 30464599
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464599 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464599 Supplier Toronto Research Chemicals Supplier No. TRCM159500250G
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material D-(+)-Malic Acid
CAS 636-61-3
Molecular Formula C4 H6 O5
Quantity 250 g
Synonym D-Malic Acid, Butanedioic acid, 2-hydroxy-, (2R)-, Butanedioic acid, hydroxy-, (2R)- (9CI), Butanedioic acid, hydroxy-, (R)-, Malic acid, D- (8CI), (2R)-2-Hydroxybutanedioic acid, (+)-Malic acid, (2R)-2-Hydroxysuccinic acid, (2R)-Malic acid, (R)-(+)-Malic acid, (R)-2-Hydroxysuccinic acid, (R)-Malic acid, (R)-Malic acid, D-(+)-2-Hydroxysuccinic acid, D-(+)-Malic acid, D-Malic acid
InChI Formula InChI=1S/C4H6O5/c5-2(4(8)9)1-3(6)7/h2,5H,1H2,(H,6,7)(H,8,9)/t2-/m1/s1
SMILES O[C@H](CC(=O)O)C(=O)O
IUPAC Name (2R)-2-hydroxybutanedioic acid
Molecular Weight (g/mol) 134.09
Formula Weight 134.0215
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.