missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Maltose monohydrate, 92+%

Product Code. 10402471 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10402471

Brand: Thermo Scientific Chemicals 329915000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6363-53-7 | C12H24O12 | 360.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H24O12
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Melting Point 130.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 31,386
Merck Index 14,5714
Specific Rotation Condition + 137.00 (20.00°C c=4,H2O,NH3)
Specific Rotation + 137.00
Solubility Information Solubility in water: 1080g/L (20°C). Other solubilities: slightly soluble in alcohol, practically insoluble in ether
Formula Weight 360.29
Percent Purity 92+%
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material D(+)-Maltose monohydrate, 90%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.