missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Maltose Monohydrate, ≥92% (Dry Basis), Ultrapure

Product Code. 15865318 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15865318 5 kg Plastic bottle 5kg
15855318 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15865318 Supplier Thermo Scientific Supplier No. J18725.A7
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D-(+)-Maltose Monohydrate, >-92% (Dry Basis), Ultrapure, Affymetrix/USB, Quantity: 5kg, Packaging: Plastic bottle, Grade: Ultrapure, Melting Point: ∼120 deg.C (decomposition), Percent Purity: >-92%, Assay Percent Notes: Dry basis, Beilstein: 5784659, CAS: 6363-53-7, EINECS Number: 200-716-5 | CAS: 6363-53-7 | C12H22O11·H2O | 360.31 g/mol

D-(+)-Maltose Monohydrate, >-92% (Dry Basis), Ultrapure, Affymetrix/USB, Quantity: 5kg, Packaging: Plastic bottle, Grade: Ultrapure, Melting Point: ∼120 deg.C (decomposition), Percent Purity: >-92%, Assay Percent Notes: Dry basis, Beilstein: 5784659, CAS: 6363-53-7, EINECS Number: 200-716-5

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6363-53-7
Molecular Formula C12H22O11·H2O
Molecular Weight (g/mol) 360.31
MDL Number MFCD00149343
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2 r,3 r,4 r,5 r-2,3,5,6-tetrahydroxy-4-2 r,3 r,4 s,5 s,6 r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O

Specifications

Melting Point ∼120°C (decomposition)
Packaging Plastic bottle
Quantity 5 kg
Beilstein 5784659
Formula Weight 360.31
Percent Purity ≥92%
Grade Ultrapure
Physical Form Crystalline Powder
Assay Dry basis
Chemical Name or Material D-(+)-Maltose Monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.