missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Maltose Monohydrate, Fisher BioReagents

Product Code. 10051403
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10051403 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10051403 Supplier Fisher BioReagents Supplier No. BP684500
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Maltose is suitable for use in cell culture systems requiring sugar additives.
TRUSTED_SUSTAINABILITY

Specifications

Alpha Vector Maltose Monohydrate
CAS 6363-53-7
Melting Point 130°C
Color Colorless
pH 5.5
Loss on Drying record%
Assay Percent Range ≥97 %
Packaging Poly Bottle
Molecular Formula C12H24O12
MDL Number MFCD00149343
Quantity 500 g
Specific Rotation 128 to 132° (+ or -)
Synonym d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
InChI Key HBDJFVFTHLOSDW-UHFFFAOYNA-N
SMILES O.OCC(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)C(O)C=O
IUPAC Name 2,3,5,6-tetrahydroxy-4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexanal hydrate
Molecular Weight (g/mol) 360.31
Formula Weight 360.29
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material D-(+)-Maltose Monohydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.