missing translation for 'onlineSavingsMsg'
Learn More

D-Mannitol, MP Biomedicals

Product Code. 11443470 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443470 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443470 Supplier MP Biomedicals Supplier No. 02102248.5
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

  • D-Mannitol used with boric acid in the manufacturing of dry electrolytic condensers for radio applications
  • In pharmacy as excipient and diluent for solids and liquids
  • In analytical chemistry for boron determinations
  • In the manufuring of mannitol hexanitrate
  • It is used as an anticaking and free-flow agent, flavoring agent, lubricant and release agent, stabilizer and thickener and nutritive sweetener in the food industry
  • Also used as a diuretic

TRUSTED_SUSTAINABILITY

Specifications

CAS 69-65-8
Melting Point 163°C to 172°C
Boiling Point 290°C to 295°C (3.5mm)(literature)
Molecular Formula C6H14O6
Linear Formula C6H14O6
MDL Number MFCD00064287
Quantity 500 g
Synonym d-mannitol, mannitol, mannite, osmitrol, manna sugar, osmofundin, cordycepic acid, mannit, mannazucker, mannidex
Solubility Information Soluble in water (1 g/5.5 mL), alcohol (1 g/83 mL), glycerol (1 g/18 mL); more soluble in hot water; soluble in pyridine, aniline, and aqueous solutions of alkalies; insoluble in ether.
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
SMILES OCC(O)C(O)C(O)C(O)CO
IUPAC Name hexane-1,2,3,4,5,6-hexol
Molecular Weight (g/mol) 182.17
PubChem CID 6251
ChEBI CHEBI:16899
Formula Weight 182.2
Percent Purity ≥96%
Physical Form Powder
Chemical Name or Material D-MANNITOL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.