missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-mannose, 100%, MP Biomedicals™

Product Code. 11483470 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483470 100 g 100g
11473470 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11483470 Supplier MP Biomedicals Supplier No. 0210225080
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6H12O6
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Color White
Density 0.35g/mL
Quantity 100 g
Formula Weight 180.2
Percent Purity 1
Physical Form Powder
Chemical Name or Material D-(+)-Mannose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.