missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Mannose, 99%

Product Code. 11469812
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11469812 500 g 500g
11449812 25 g 25g
11459812 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11469812 Supplier Thermo Scientific Chemicals Supplier No. A10842.36
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3458-28-4 | C6H12O6

D-mannose acts as an alternative energy source utilized by brain. It is used in the study of assessing the synthesis of amphiphilic glycopolymers and investigating early detection of bronchiolitis obliterans.It may be useful in the treatment of urinary tract infections (UTIs) and carbohydrate-deficient glycoprotein syndrome type 1b. It is also useful in the formation of glycan structure and glycosylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-mannose acts as an alternative energy source utilized by brain. It is used in the study of assessing the synthesis of amphiphilic glycopolymers and investigating early detection of bronchiolitis obliterans.It may be useful in the
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
MDL Number MFCD00064122
Synonym 2s,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanal, polymannose, aldehydo-d-mannose, poly mannose, mannose homopolymer, unii-pha4727wtp, aldehydo-d-manno-hexose, pha4727wtp, mannose, d, d-mannose polymers

Specifications

Melting Point 127°C to 133°C
Density 1.54
Molecular Formula C6H12O6
Quantity 500 g
Beilstein 1564373
Sensitivity Hygroscopic
Merck Index 14,5747
Solubility Information Soluble in water.
Optical Rotation +13.8° (c=10 in water, 24h)
Formula Weight 180.16
Percent Purity 99%
Chemical Name or Material D-(+)-Mannose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.