missing translation for 'onlineSavingsMsg'
Learn More

D-Mannose, TRC

Product Code. 30009828
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009828 10 g 10g
30005440 100 g 100g
30009349 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009828 Supplier Toronto Research Chemicals Supplier No. TRCM16800010G
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3458-28-4
Molecular Formula C6 H12 O6
Molecular Weight (g/mol) 180.16
Synonym D-Mannose, Mannose, D- (8CI), (+)-Mannose, Carubinose, D(+)-Mannose, Mannose, NSC 26247, Seminose
IUPAC Name (3S,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol
SMILES OC[C@H]1OC(O)[C@@H](O)[C@@H](O)[C@@H]1O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5+,6?/m1/s1
Formula Weight 180.0634
Chemical Name or Material D-(+)-Mannose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.