missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Melezitose monohydrate, 99+%

Product Code. 10696222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10696222 25 g Glass bottle 25g
10082990 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10696222 Supplier Thermo Scientific Chemicals Supplier No. 225640250
 Restricted Item
Estimated Shipment: 10-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D(+)-Melezitose monohydrate, CAS # 597-12-6, is the D-form hydrate of melezitose, the trisaccharide.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Sap-feeding insects such as ants and aphids produce melezitose to maintain osmotic balance

• Melezitose is produced by some plants as an insect attractant and acts as a metabolite

Applications

• Melezitose can be hydrolyzed to glucose and turanose through alpha-glucosidase activity and can be used for the study of enzyme activity

• Melezitose can be used to distinguish certain strains of bacteria; characterization of phenotype based on the presence or absence of certain sugars assists identification

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10030-67-8
Molecular Formula C18H36O18
Molecular Weight (g/mol) 540.468
MDL Number MFCD00149448
InChI Key LNVIPYYEBMNJIL-ZWELICPFSA-N
Synonym melezitose dihydrate, d-melezitosedihydrate, unii-d12c11k0m0, d-melezitose dihydrate, melezitose dihydrate mi, melezitose dihydrate, +, alpha-d-glucopyranoside, o-alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl, dihydrate, o-alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl-alpha-d-glucopyranoside dihydrate, alpha-d-glucopyranosyl-1->3-beta-d-fructofuranosyl alpha-d-glucopyranoside-water 1/2, 2r,3r,4s,5s,6r-2-2s,3s,4r,5r-4-hydroxy-2,5-bis hydroxymethyl-2-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy oxolan-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol dihydrate
PubChem CID 67660249
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2S,3S,4R,5R)-4-hydroxy-2,5-bis(hydroxymethyl)-2-[(2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxyoxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol;dihydrate
SMILES C(C1C(C(C(C(O1)OC2C(C(OC2(CO)OC3C(C(C(C(O3)CO)O)O)O)CO)O)O)O)O)O.O.O

Specifications

Melting Point 160.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Quantity 25 g
Beilstein 31, 466
Merck Index 14, 5819
Specific Rotation Condition +88° (20°C c=4,H2O)
Specific Rotation + 88.00
Formula Weight 522.45
Percent Purity ≥99%
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Melezitose monohydrate, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.