missing translation for 'onlineSavingsMsg'
Learn More

α-D-(+)-Melibiose Monohydrate, MP Biomedicals™

Product Code. 11483480 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
Unit Size:
100g
10g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11483480 5 g 5g
11493480 10 g 10g
11403490 25 g 25g
11413490 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11483480 Supplier MP Biomedicals Supplier No. 0210225605
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

α-D-(+)-Melibiose Monohydrate used to improve the preparation of benzoylated ethyl 1-thioglycosidesto and to study amorphous disaccharides.

  • Melibiose has been used in a study to improve the preparation of benzoylated ethyl 1-thioglycosides.
  • It has also been used in a study to investigate the effect of water plasticization on the molecular mobility and crystallization tendency of amorphous disaccharides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 585-99-9
Molecular Formula C12H22O11·H2O
Synonym 6-O-α-D-galactopyranosyl-D-glucose, α-D-melibiose hydrate

Specifications

Color White
Quantity 5 g
Optical Rotation +135 ± 10° (c=4, water)
Formula Weight 360.3
Percent Purity 99.9%
Physical Form Powder
Chemical Name or Material α-D-(+)-Melibiose monohydrate

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.