missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Methionine-3,3,4,4-d4, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | D-Methionine-d4 |
| Molecular Formula | C5 D4 H7 N O2 S |
| Quantity | 0.05 g |
| Synonym | D-Methionine-3,3,4,4-d4, D-Methionine-d4, (2 R)-2-Amino-4-methylsulfanylbutanoic-3,3,4,4-d4 acid, (R)-2-Amino-4-(methylthio)butanoic-3,3,4,4-d4 acid, S-Methyl-D-homocysteine-d4 |
| InChI Formula | InChI=1 S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m1/s1/i2D2,3D2 |
| SMILES | [2 H]C([2 H])(SC)C([2 H])([2 H])[C@@H](N)C(=O)O |
| IUPAC Name | (2 R)-2-amino-3,3,4,4-tetradeuterio-4-methylsulfanylbutanoic acid |
| Molecular Weight (g/mol) | 153.236 |
| Formula Weight | 153.076 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction