missing translation for 'onlineSavingsMsg'
Learn More

alpha-D-Methylglucoside, 99%

Product Code. p-162798 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10092070

Brand: Thermo Scientific Acros 173841000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 97-30-3 | C7H14O6 | 194.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-30-3
Molecular Formula C7H14O6
Molecular Weight (g/mol) 194.18
MDL Number MFCD00064086
InChI Key HOVAGTYPODGVJG-IECVIRLLSA-N
Synonym methyl alpha-d-glucopyranoside, methyl alpha-d-glucoside, alpha-methylglucoside, alpha-methyl-d-glucoside, alpha-methyl d-glucose ether, methyl-alpha-d-glucopyranoside, alpha-d-methylglucoside, me alpha-glc, alpha-d-methyl glucoside, unii-qcf122nf3r
PubChem CID 7568545
IUPAC Name (2R,3R,4R,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
SMILES COC1C(C(C(C(O1)CO)O)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 169.0°C to 171.0°C
Color White
Boiling Point 200.0°C (0.2 mmHg)
Loss on Drying 0.1% max. (60 to 70°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 31, 179
Merck Index 15, 6152
Specific Rotation Condition +158.50° (20°C c=10,water)
Specific Rotation + 158.50
Solubility Information Solubility in water: 108g/100mL (20 c). Other solubilities: practically insoluble in ether,5.2g/100g methanol (20 c),7.3g/100g alcohol 80% (17 c)
Formula Weight 194.18
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material α-D-Methylglucoside, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.