missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ D-myo-Inositol 1,4,5-trisphosphate, hexapotassium salt

Product Code. 18014192
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18014192 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18014192 Supplier R&D Systems™ Supplier No. 1482/1
  • ONLINE EXCLUSIVE PRICE
    283.40 €
    253.05 € / 1mg
    Save 30.35 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Ca2+ mobilizing second messenger

Important second messenger involved in Ca2+ mobilization from intracellular stores (EC50 = 0.1 μM). Formed from the enzymatic hydrolysis of phosphatidyl inositol-4,5-bisphosphate.
TRUSTED_SUSTAINABILITY

Specifications

CAS 103476-24-0
Quantity 1 mg
Product Line Tocris
Molecular Formula C6H9K6O15P3
Formula Weight 648.64
Physical Form Solid
Molecular Weight (g/mol) 648.634
Chemical Name or Material D-myo-Inositol 1,4,5-trisphosphate, hexapotassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.